Learn More
Medchemexpress LLC Ac-rC phosphoramidite | 121058-88-6 | 98.9% | 902.10 g/mol | C47H64N5O9PSi | 1 G
SDP
Catalog No. 5000210747
Encompass_Preferred
Ac-rC phosphoramidite is a protected cytidine phosphoramidite reagent designed for solid-phase oligoribonucleotide synthesis, including preparation of phosphorodithioate (PS2-RNA) modified sequences. It is supplied as a solid or as DMSO solutions for immediate use in automated or manual RNA synthesis workflows.
  • Used for oligoribonucleotide and PS2-RNA synthesis.
  • Contains 5'-DMT and acetyl-protected cytidine base.
  • High purity (98.9%) suitable for synthesis applications.
  • Available as solid and 10 mM DMSO solutions for convenience.
  • White to off-white solid appearance for easy visual inspection.
  • Store protected from light at low temperatures; in solution keep frozen for extended stability.
Catalog No. 50-002-10747 Supplier Medchemexpress LLC Supplier No. HYW0423571G
Ac-rC phosphoramidite is a protected cytidine phosphoramidite reagent designed for solid-phase oligoribonucleotide synthesis, including preparation of phosphorodithioate (PS2-RNA) modified sequences. It is supplied as a solid or as DMSO solutions for immediate use in automated or manual RNA synthesis workflows.
  • Used for oligoribonucleotide and PS2-RNA synthesis.
  • Contains 5'-DMT and acetyl-protected cytidine base.
  • High purity (98.9%) suitable for synthesis applications.
  • Available as solid and 10 mM DMSO solutions for convenience.
  • White to off-white solid appearance for easy visual inspection.
  • Store protected from light at low temperatures; in solution keep frozen for extended stability.