Learn More
Enzo Life Sciences Ac-REQD-pNA (5mg)
SDP
Supplier:  Enzo Life Sciences BMLP4420005
Encompass_Preferred
Colorimetric caspase substrate. Sequence corresponds to the prodomain of procaspase-8 cleavage site, residues 213-216, a putative caspase-10 target. Selective caspase-1 substrate at concentrations of 0.2-2 mM. Alternative name: Caspase 1 substrate (chromogenic). Purity: ≥95%. Long Term Storage: -80°C.
Catalog No. 50-201-1351