Learn More
CPC Scientific Ac-Arg-Gly-Lys(Ac)-AMC (trifluoroacetate salt) 5MG
SDP
Catalog No. 502109432
Encompass
This peptide is the fluorogenic substrate for assaying histone deacetylase (HDAC) activity in a two-step enzymatic reaction. The assay consists of the initial lysine deacetylation by HDAC followed by the release of the fluorescent group by trypsin.ONE-LETTER SEQUENCE: Ac-RGK(Ac)-AmcMOLECULAR FORMULA: C28H40N8O7MOLECULAR WEIGHT:600.68STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [660846-97-9], RESEARCH AREA: MiscellaneousREFERENCES: D.Wegener et al., Chem. Biol., 10, 61 (2003)
Catalog No. 50-210-9432 Supplier CPC Scientific Supplier No. HIST006A
This peptide is the fluorogenic substrate for assaying histone deacetylase (HDAC) activity in a two-step enzymatic reaction. The assay consists of the initial lysine deacetylation by HDAC followed by the release of the fluorescent group by trypsin.ONE-LETTER SEQUENCE: Ac-RGK(Ac)-AmcMOLECULAR FORMULA: C28H40N8O7MOLECULAR WEIGHT:600.68STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [660846-97-9], RESEARCH AREA: MiscellaneousREFERENCES: D.Wegener et al., Chem. Biol., 10, 61 (2003)