Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Checkers AC Series Aviation Wheel Chocks
GREENER_CHOICE

Catalog No. 19088481 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Lime Green
Yellow
Includes:
Locking Rope
Loop Rope
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Includes
19-088-481 Yellow Locking Rope
19-088-480 Lime Green Locking Rope
19-088-479 Lime Green Loop Rope
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-088-481 Supplier Checkers Supplier No. AC6820LR
Add to Cart
Add to Cart

Lightweight polyurethane aviation wheel chocks are ideal for private, commercial, cargo, and military aircraft.

  • Checkers™ Aviation Wheel Chocks are made of highly durable, lightweight polyurethane
  • AC Chocks last longer than wood chocks, never need painting and eliminate Foreign Object Damage threats for aircraft
  • The AC Series is ideal for private, commercial, cargo and military aircraft

Specifications

For Use With (Equipment) Private, Commercial, Cargo, and Military Aircrafts
Color Yellow
Product Type Wheel Chock
Dimensions (L x W x H) 20 x 8 x 7 in.
Includes Locking Rope
Material Polyurethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.