Learn More
Medchemexpress LLC Ac-Trp-Leu-Ala-AMC | 1104011-59-7 | 99.8% | 587.7 g/mol | C32H37N5O6 | 10MG
SDP
Catalog No. 5000236325
Encompass_Preferred
Ac-WLA-AMC (Ac-Trp-Leu-Ala-AMC; CAS 1104011-59-7) is an acetylated tripeptidyl fluorogenic substrate for measuring the chymotrypsin-like activity of the 20S proteasome β5 (β5c) subunit. Enzymatic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal used in proteasome activity assays and inhibitor screening.
  • Measures chymotrypsin-like proteasome β5 activity.
  • Releases 7-amino-4-methylcoumarin (AMC) for sensitive fluorescence detection.
  • Compatible with kinetic and end-point fluorescence assays.
  • Supplied as a solid peptide substrate suitable for biochemical assays.
  • High purity (99.8%) appropriate for research use.
Catalog No. 50-002-36325 Supplier Medchemexpress LLC Supplier No. HY12305110MG
Ac-WLA-AMC (Ac-Trp-Leu-Ala-AMC; CAS 1104011-59-7) is an acetylated tripeptidyl fluorogenic substrate for measuring the chymotrypsin-like activity of the 20S proteasome β5 (β5c) subunit. Enzymatic cleavage releases 7-amino-4-methylcoumarin (AMC), producing a fluorescent signal used in proteasome activity assays and inhibitor screening.
  • Measures chymotrypsin-like proteasome β5 activity.
  • Releases 7-amino-4-methylcoumarin (AMC) for sensitive fluorescence detection.
  • Compatible with kinetic and end-point fluorescence assays.
  • Supplied as a solid peptide substrate suitable for biochemical assays.
  • High purity (99.8%) appropriate for research use.