Learn More
Enzo Life Sciences Acetylated Alpha Tubulin (K40) Recombinant monoclonal antibody (RM318) (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS6760100
Encompass_Preferred
Clone: RM318. Isotype: IgG. Immunogen: Synthetic acetyl-peptide corresponding to the K40 region of human alpha tubulin. Source: Purified from animal origin-free supernatant. Species reactivity: Human. Applications: ICC, IF, WB. Purity: Protein A affinity purified. Formulation: Liquid. In 1x PBS containing 50% glycerol, 1% BSA and 0.09% sodium azide.
Catalog No. 50-201-3204