Learn More
CPC Scientific Ac-Tyr-Val-Ala-Asp-AFC 1MG
SDP
Supplier:  CPC Scientific CASP009A
Encompass
Halogen substituents that improve membrane permeability when incorporated into substrate.SEQUENCE: Ac-Tyr-Val-Ala-Asp-AFCONE-LETTER SEQUENCE: Ac-YVAD-AFCMOLECULAR FORMULA: C33H36F3N5O10MOLECULAR WEIGHT: 719.67STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [219137-85-6]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:B. Zhivotovsky et al., Exp. Cell Res., 221, 404 (1995)J. Xiang et al., Proc. Natl. Acad. Sci. USA , 93, 14559 (1996)H. Ohnimus, Personal communication (1997)SKU(s): CASP-009A
Catalog No. 50-210-8937