Learn More
Medchemexpress LLC Ac-Tyr-Val-Ala-Asp-AMC | 149231-65-2 | 665.69 | 1 MG
SDP
Catalog No. 5000400390
Encompass_Preferred
Ac-YVAD-AMC is a potent caspase inhibitor that effectively suppresses bacteria-induced cell death, specifically within hypersensitive response (HR) cells. This compound is valuable in research for studying cellular apoptosis and inflammatory processes.
  • Inhibits caspase activity
  • Suppresses bacteria-induced cell death in HR cells
  • Useful for apoptosis research
  • Supports studies on inflammatory pathways
Catalog No. 50-004-00390 Supplier Medchemexpress LLC Supplier No. HYP27171MG
Ac-YVAD-AMC is a potent caspase inhibitor that effectively suppresses bacteria-induced cell death, specifically within hypersensitive response (HR) cells. This compound is valuable in research for studying cellular apoptosis and inflammatory processes.
  • Inhibits caspase activity
  • Suppresses bacteria-induced cell death in HR cells
  • Useful for apoptosis research
  • Supports studies on inflammatory pathways