Learn More
Bachem Ac-Tyr-Val-Ala-Asp-chloromethylketone
SDP
Supplier:  Bachem 4018838.0005
Encompass_Preferred
Ac-Tyr-Val-Ala-Asp-chloromethylketone, 178603-78-6, C24H33ClN4O8, Mr 541.01, 5mg. Ac-YVAD-CMK. Cell-permeable, irreversible inhibitor of caspase-1 (ICE). N.Rouquet et al. showed that Ac-YVAD-CMK is a potent therapeutic agent against in vivo liver apoptosis in mice.
Catalog No. 50-260-537