Learn More
Enzo Life Sciences Ac-VAD-AFC (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260109M005
Encompass_Preferred
Fluorogenic substrate for all caspases. Alternative name: Caspase substrate (fluorogenic) (all caspases). Formula: C24H27F3N4O8. MW: 556.5. Solubility: Soluble in DMF or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8321