Learn More
CPC Scientific Ac-Val-Ala-Asp-CHO 1MG
SDP
Supplier:  CPC Scientific CASP016A
Encompass
SEQUENCE: Ac-Val-Ala-Asp-CHOONE-LETTER SEQUENCE: Ac-VAD-CHOMOLECULAR FORMULA: C14H23N3O6MOLECULAR WEIGHT: 329.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [147837-52-3]SYNONYMS: Caspase Inhibitor IIRESEARCH AREA: ApoptosisREFERENCES:SKU(s): CASP-016A
Catalog No. 50-210-8949