Learn More
CPC Scientific Ac-Val-Glu-His-Asp-AFC (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CASP018A
Encompass
ONE-LETTER SEQUENCE: Ac-VEHD-AFCMOLECULAR FORMULA: C32H36N7O11F3MOLECULAR WEIGHT:751.25STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [1135418-22-2], RESEARCH AREA: Apoptosis
Catalog No. 50-210-8953