Learn More
Enzo Life Sciences Ac-VDVAD-CHO (5mg). CAS: 194022-51-0
SDP
Catalog No. 502008270
Encompass_Preferred
Potent, reversible aldehyde inhibitor of caspase-2 (Ki=3.5nM). Preferred cleavage sequence for caspase-2. Alternative name: Caspase 2 inhibitor (aldehyde). Formula: C23H37N5O10. MW: 543.6. Purity: ≥94% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO (100mM, 54.4mg/ml) or buffer (pH 7-7.5). Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8270 Supplier Enzo Life Sciences Supplier No. ALX260058M005
Potent, reversible aldehyde inhibitor of caspase-2 (Ki=3.5nM). Preferred cleavage sequence for caspase-2. Alternative name: Caspase 2 inhibitor (aldehyde). Formula: C23H37N5O10. MW: 543.6. Purity: ≥94% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO (100mM, 54.4mg/ml) or buffer (pH 7-7.5). Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. Handling: Keep cool and dry.