Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-VDVAD-CHO (TFA) | 00-00-0 | 99.2% | 657.59 g·mol⁻¹ | C25H38F3N5O12 | 5 MG
SDP

Catalog No. 5000222476
Encompass_Preferred

Ac-VDVAD-CHO (TFA) is a peptide aldehyde inhibitor used in biochemical and cell-based research to block caspase-mediated proteolysis and investigate apoptosis pathways.

  • Selective caspase-2 and caspase-3 inhibition (IC50 46 nM and 15 nM).
  • TFA salt, supplied as a white to off-white solid.
  • High purity (99.2%).
  • Molecular weight 657.59 g·mol⁻¹; formula C25H38F3N5O12.
  • Recommended storage: powder at -80°C (2 years) or -20°C (1 year).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-22476 Supplier Medchemexpress LLC Supplier No. HY134567A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-VDVAD-CHO (TFA) is a peptide aldehyde inhibitor used in biochemical and cell-based research to block caspase-mediated proteolysis and investigate apoptosis pathways.

  • Selective caspase-2 and caspase-3 inhibition (IC50 46 nM and 15 nM).
  • TFA salt, supplied as a white to off-white solid.
  • High purity (99.2%).
  • Molecular weight 657.59 g·mol⁻¹; formula C25H38F3N5O12.
  • Recommended storage: powder at -80°C (2 years) or -20°C (1 year).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.