Learn More
Medchemexpress LLC Ac-VDVAD-CHO (trifluoroacetate salt) | 00-00-0 | 99.2% | C25H38F3N5O12 | 10MG
SDP
Catalog No. 5000259244
Encompass_Preferred
Ac-VDVAD-CHO (TFA) is a peptide aldehyde inhibitor of caspase-2 and caspase-3 supplied as the trifluoroacetate salt for research use. It exhibits potent activity at low nanomolar concentrations (caspase-2 IC50 ≈ 46 nM; caspase-3 IC50 ≈ 15 nM), has molecular formula C25H38F3N5O12, and is supplied with high purity appropriate for biochemical and apoptosis assays.
  • Selective inhibitor of caspase-2 and caspase-3.
  • Potent activity in the low nanomolar range.
  • Suitable for apoptosis and protease activity studies.
  • High purity for reliable experimental results.
  • Provided as a trifluoroacetate salt for improved handling.
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-59244 Supplier Medchemexpress LLC Supplier No. HY134567A10MG
Ac-VDVAD-CHO (TFA) is a peptide aldehyde inhibitor of caspase-2 and caspase-3 supplied as the trifluoroacetate salt for research use. It exhibits potent activity at low nanomolar concentrations (caspase-2 IC50 ≈ 46 nM; caspase-3 IC50 ≈ 15 nM), has molecular formula C25H38F3N5O12, and is supplied with high purity appropriate for biochemical and apoptosis assays.
  • Selective inhibitor of caspase-2 and caspase-3.
  • Potent activity in the low nanomolar range.
  • Suitable for apoptosis and protease activity studies.
  • High purity for reliable experimental results.
  • Provided as a trifluoroacetate salt for improved handling.
  • Available in small research pack sizes for laboratory use.