Learn More
Enzo Life Sciences Ac-VDVAD-pNA (1mg). CAS: 189684-53-5
SDP
Supplier:  Enzo Life Sciences ALX260059M001
Encompass_Preferred
Caspase-2 substrate. Chromogenic substrate for caspase-2. Preferred substrate for caspase-2 (Km=53μM) Alternative name: Caspase 2 substrate (chromogenic). Formula: C29H41N7O12. MW: 679.7. Purity: ≥98% (HPLC). Formulation: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in DMSO (100mM). Dilute stock solution with distilled water or buffer. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8271