Learn More
Medchemexpress LLC Ac-VEID-AFC | 210345-01-0 | 99.3% | 727.68 | 1 MG
SDP
Catalog No. 5000396160
Encompass_Preferred
Ac-VEID-AFC is a caspase-6 fluorogenic substrate used in caspase 6 activity assays. This product is a solid, appearing white to off-white, and is shipped at room temperature. It is utilized in various disease areas including cancer, cancer targeted therapy, cancer metabolism and metastasis, apoptosis, and caspase research.
  • Used in caspase 6 activity assays
  • White to off-white solid appearance
  • Ships at room temperature in continental US
  • Related to peptides, enzyme substrates, and various disease areas
Catalog No. 50-003-96160 Supplier Medchemexpress LLC Supplier No. HYP59831MG
Ac-VEID-AFC is a caspase-6 fluorogenic substrate used in caspase 6 activity assays. This product is a solid, appearing white to off-white, and is shipped at room temperature. It is utilized in various disease areas including cancer, cancer targeted therapy, cancer metabolism and metastasis, apoptosis, and caspase research.
  • Used in caspase 6 activity assays
  • White to off-white solid appearance
  • Ships at room temperature in continental US
  • Related to peptides, enzyme substrates, and various disease areas