Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-VEID-AFC | 210345-01-0 | 99.3% | 727.68 | 10 MG
SDP

Catalog No. 5000396193
Encompass_Preferred

Ac-VEID-AFC is a caspase-6 fluorogenic substrate, used in caspase 6 activity assays. It is for research use only.

  • Used in caspase 6 activity assays
  • For research use only
  • Store as powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Requires sealed storage, away from moisture and light

Catalog No. 50-003-96193 Supplier Medchemexpress LLC Supplier No. HYP598310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ac-VEID-AFC is a caspase-6 fluorogenic substrate, used in caspase 6 activity assays. It is for research use only.

  • Used in caspase 6 activity assays
  • For research use only
  • Store as powder at -80°C for 2 years or -20°C for 1 year
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Requires sealed storage, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.