Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-VEID-AFC | 210345-01-0 | 99.3% | 727.68 | 5 MG
SDP

Catalog No. 5000397183
Encompass_Preferred

Ac-VEID-AFC is a fluorogenic substrate for caspase-6, widely utilized in caspase 6 activity assays.

  • Functions as a fluorogenic substrate for caspase-6.
  • Suitable for use in caspase 6 activity assays.
  • High purity (99.3%).
  • Molecular weight of 727.68.
  • Appears as a white to off-white solid.
  • Soluble in water.

Catalog No. 50-003-97183 Supplier Medchemexpress LLC Supplier No. HYP59835MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-VEID-AFC is a fluorogenic substrate for caspase-6, widely utilized in caspase 6 activity assays.

  • Functions as a fluorogenic substrate for caspase-6.
  • Suitable for use in caspase 6 activity assays.
  • High purity (99.3%).
  • Molecular weight of 727.68.
  • Appears as a white to off-white solid.
  • Soluble in water.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.