Learn More
Enzo Life Sciences Ac-VEID-AFC (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260111M005
Encompass_Preferred
Caspase-6 inhibitor. Fluorogenic substrate for caspase-6 (Km=30μM; Kcat/Km=168000 M-1sec-1) and related cysteine proteases. Sequence is based on lamin A site of cleavage. Alternative name: Caspase 6 substrate (fluorogenic). Formula: C32H40F3N5011. MW: 727.7. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or dimethyl formamide. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8322