Learn More
Cayman Chemical Ac-VEID-AMC (ammonium salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502969954
Encompass_Preferred
A fluorogenic substrate that can be cleaved by caspase-6 and related proteases including caspase-8
Catalog No. 50-296-9954 Supplier Cayman Chemical Supplier No. 149891 MG
A fluorogenic substrate that can be cleaved by caspase-6 and related proteases including caspase-8