Learn More
Medchemexpress LLC Ac-VEID-CHO (trifluoroacetate salt) | 00-00-0 | 99.8% | 614.57 Da | C24H37F3N4O11 | 1 MG
SDP
Catalog No. 5000241902
Encompass_Preferred
Ac-VEID-CHO (TFA) is a peptide-derived reversible caspase inhibitor intended for in vitro research into apoptosis and neurodegenerative pathways. It is supplied as a trifluoroacetate salt and demonstrates potent inhibition of several executioner caspases in biochemical assays. Not for human or clinical use.
  • Reversible caspase inhibitor targeting caspase-6, caspase-3, and caspase-7.
  • Reported IC50 values in the low nanomolar range for primary targets.
  • High purity suitable for biochemical assays (99.8%).
  • Available in small mg-scale quantities for research use.
  • Limited cellular activity due to poor cellular accumulation.
Catalog No. 50-002-41902 Supplier Medchemexpress LLC Supplier No. HY108312A1MG
Ac-VEID-CHO (TFA) is a peptide-derived reversible caspase inhibitor intended for in vitro research into apoptosis and neurodegenerative pathways. It is supplied as a trifluoroacetate salt and demonstrates potent inhibition of several executioner caspases in biochemical assays. Not for human or clinical use.
  • Reversible caspase inhibitor targeting caspase-6, caspase-3, and caspase-7.
  • Reported IC50 values in the low nanomolar range for primary targets.
  • High purity suitable for biochemical assays (99.8%).
  • Available in small mg-scale quantities for research use.
  • Limited cellular activity due to poor cellular accumulation.