Learn More
Cole-Parmer Extech DVA30 AC Volt / Current Detector Non-Contact Ac
Supplier:  Cole-Parmer 2000527
Encompass
Detects and pinpoints wide range voltage and current with sensitivity adjust
  • Current function detects live circuits through shielded wires, conduit and metal circuit breaker or junction boxes where voltage detectors cannot work
  • Non-contact detection of AC voltage from 12VAC to 600VAC; detects AC Current from 200mA at 0.2" and larger current at greater distances
  • Sensitivity adjustment increases or reduces sensor trigger threshold
  • Dimensions / Wt: 7.6 x 1.2 x 0.9" (192 x 31 x 24mm) / 2.1 oz (60g)
  • Audible and visual indicator of current or voltage
Catalog No. 50-992-598