Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-vrpr-amc tfa | 919515-51-8 | 99.6% | 953.88 | 5 MG
SDP

Catalog No. 5000396819
Encompass_Preferred

Ac-VRPR-AMC TFA is a fluorogenic metacaspase substrate. It can be used to assay metacaspase activity. This product is for research use only.

  • Fluorogenic metacaspase substrate
  • Suitable for assaying metacaspase activity
  • Research use only
  • Molecular weight: 953.88
  • Purity: 99.6%
  • White to off-white solid appearance
  • Soluble in water at 100 mg/mL or more

Catalog No. 50-003-96819 Supplier Medchemexpress LLC Supplier No. HYP4449A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-VRPR-AMC TFA is a fluorogenic metacaspase substrate. It can be used to assay metacaspase activity. This product is for research use only.

  • Fluorogenic metacaspase substrate
  • Suitable for assaying metacaspase activity
  • Research use only
  • Molecular weight: 953.88
  • Purity: 99.6%
  • White to off-white solid appearance
  • Soluble in water at 100 mg/mL or more

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.