Learn More
Enzo Life Sciences Ac-WEHD-AFC (10mg)
SDP
Catalog No. 502008329
Encompass_Preferred
Fluorogenic substrate for caspase-1, -4, -5 and -14. Alternative name: Caspase 1 substrate (fluorogenic), Caspase 5 substrate (fluorogenic). Formula: C38H37F3N8O11. MW: 838.8. Purity: ≥98% (HPLC). Appearance: White to yellow powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8329 Supplier Enzo Life Sciences Supplier No. ALX260117M010
Add to Cart
Add to Cart
Fluorogenic substrate for caspase-1, -4, -5 and -14. Alternative name: Caspase 1 substrate (fluorogenic), Caspase 5 substrate (fluorogenic). Formula: C38H37F3N8O11. MW: 838.8. Purity: ≥98% (HPLC). Appearance: White to yellow powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Handling: Keep cool and dry.