Learn More
Enzo Life Sciences Ac-WEHD-AFC (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260117M005
Encompass_Preferred
Substrate for caspase-1, -4-, -5 and -14. Fluorogenic substrate for caspase-1, -4, -5 and -14. Alternative name: Caspase 1 substrate (fluorogenic), Caspase 5 substrate (fluorogenic). Formula: C38H37F3N8O11. MW: 838.8. Purity: ≥98% (HPLC). Appearance: White to yellow powder. Peptide content: 70-90%. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Solution in DMSO is stable for 6-8 months when stored at -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8328