Learn More
Abcam Ac-WEHD-AFC, 25MG
Catalog No. 502205508
Encompass_Preferred
MW 838.7 Da, Purity >97%. Ac-WEHD-AFC is a useful fluorogenic substrate for group I caspases (caspase-1, -4, and -5). Caspase activity can be quantified by fluorescent detection of free AFC that has an excitation maximum at 400 nm and emission maximum at 505 nm.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5508 Supplier Abcam Supplier No. AB28537525MG
MW 838.7 Da, Purity >97%. Ac-WEHD-AFC is a useful fluorogenic substrate for group I caspases (caspase-1, -4, and -5). Caspase activity can be quantified by fluorescent detection of free AFC that has an excitation maximum at 400 nm and emission maximum at 505 nm.
The product is subject to the following: Abcam Restricted Use Statement