Learn More
Medchemexpress LLC Ac-WEHD-AFC (TFA) | 98.84% | 952.77 | 10 MG
SDP
Catalog No. 5000422942
Encompass_Preferred
Ac-WEHD-AFC TFA is a fluorogenic caspase-1 substrate that can be used to measure caspase-1 fluorogenic activity. It is applicable for research related to tumors and inflammation.
  • Fluorogenic caspase-1 substrate
  • Measures caspase-1 fluorogenic activity
  • Applicable for research related to tumors and inflammation
  • For research use only
Catalog No. 50-004-22942 Supplier Medchemexpress LLC Supplier No. HYP2617A10MG
Ac-WEHD-AFC TFA is a fluorogenic caspase-1 substrate that can be used to measure caspase-1 fluorogenic activity. It is applicable for research related to tumors and inflammation.
  • Fluorogenic caspase-1 substrate
  • Measures caspase-1 fluorogenic activity
  • Applicable for research related to tumors and inflammation
  • For research use only