Learn More
Enzo Life Sciences Ac-WEHD-CHO (5mg)
SDP
Catalog No. 502008266
Encompass_Preferred
Reversible, cell permeable inhibitor of caspase-1 and -5. Also inhibits caspase-4 and -14. Alternative name: Caspase 1 inhibitor (aldehyde), Caspase 5 inhibitor (aldehyde). Formula: C28H33N7O9. MW: 611.6. Purity: ≥90% (HPLC). Appearance: White to off-white powder. Formulation: TFA salt. Solubility: Soluble in water (10mg/ml). Long Term Storage: -20°C. Use/Stability: Solution is stable for 1 month when stored at -20°C and protected from light. Handling: Keep cool and dry.
Catalog No. 50-200-8266 Supplier Enzo Life Sciences Supplier No. ALX260055M005
Reversible, cell permeable inhibitor of caspase-1 and -5. Also inhibits caspase-4 and -14. Alternative name: Caspase 1 inhibitor (aldehyde), Caspase 5 inhibitor (aldehyde). Formula: C28H33N7O9. MW: 611.6. Purity: ≥90% (HPLC). Appearance: White to off-white powder. Formulation: TFA salt. Solubility: Soluble in water (10mg/ml). Long Term Storage: -20°C. Use/Stability: Solution is stable for 1 month when stored at -20°C and protected from light. Handling: Keep cool and dry.