Learn More
Bachem Ac-Trp-Glu-His-Asp-aldehyde (pseudo acid)
SDP
Supplier:  Bachem 4027878.0025
Encompass_Preferred
Ac-Trp-Glu-His-Asp-aldehyde (pseudo acid), 189275-71-6, C28H33N7O9, Mr 611.61, 25mg. Ac-WEHD-CHO. Ac-WEHD-CHO, a very potent reversible inhibitor of caspase-1 (ICE). It bears the optimal tetrapeptide recognition motif for this enzyme. With a Ki of 56 pM it belongs to the most potent reversible, peptide-based inhibitors described for any protease. For caspase-8 a Ki of 21.1 nM has been reported.
Catalog No. 50-194-6682