Learn More
Medchemexpress LLC Ac-yevd-amc | 1631716-11-4 | 99.59% | 723.73 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP101421MG
Encompass_Preferred
Ac-YEVD-AMC is a fluorescent substrate of caspase-1/11. It releases amino-4-methylcoumarin (AMC) for fluorescence detection when treated with cell lysate.
  • It functions as a fluorescent substrate for caspase-1/11.
  • Its mechanism involves the release of amino-4-methylcoumarin (AMC) for fluorescence detection.
  • It is intended for research use only.
Catalog No. 50-004-23240