Learn More
Medchemexpress LLC Ac-YEVD-AMC | 1631716-11-4 | 99.6% | 723.73 | 5 MG
SDP
Catalog No. 5000422507
Encompass_Preferred
Ac-YEVD-AMC is a fluorescent substrate of caspase-1/11 that releases amino-4-methylcoumarin (AMC) for fluorescence detection when treated with cell lysate. This product is for research use only.
  • Fluorescent substrate of caspase-1/11
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection
  • For research use only
Catalog No. 50-004-22507 Supplier Medchemexpress LLC Supplier No. HYP101425MG
Ac-YEVD-AMC is a fluorescent substrate of caspase-1/11 that releases amino-4-methylcoumarin (AMC) for fluorescence detection when treated with cell lysate. This product is for research use only.
  • Fluorescent substrate of caspase-1/11
  • Releases amino-4-methylcoumarin (AMC) for fluorescence detection
  • For research use only