Learn More
Cayman Chemical Ac-YVAD-AFC 10mg
SDP
Catalog No. 502979783
Encompass_Preferred
A fluorogenic substrate whose amino acids YVAD have been shown to be a preferred cleavage site for caspase-1 and -4
Catalog No. 50-297-9783 Supplier Cayman Chemical Supplier No. 1759110 MG
A fluorogenic substrate whose amino acids YVAD have been shown to be a preferred cleavage site for caspase-1 and -4