Learn More
Enzo Life Sciences Ac-YVAD-AFC (10mg)
SDP
Catalog No. 502008320
Encompass_Preferred
Substrate for caspase-1 and -4. Fluorogenic substrate for caspase-1 and -4. Alternative name: Caspase 1 substrate (fluorogenic), Caspase 4 substrate (fluorogenic). Formula: C33H36F3N5O10. MW: 719.7. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Solid product is stable for 1 year in desiccator when stored at room temperature. Stock solutions are stable for 6 – 8 months when stored at –20°C. Keep sealed after removing from the freezer until its temperature equilibrates with room temperature. Handling: Keep cool and dry.
Catalog No. 50-200-8320 Supplier Enzo Life Sciences Supplier No. ALX260108M010
Substrate for caspase-1 and -4. Fluorogenic substrate for caspase-1 and -4. Alternative name: Caspase 1 substrate (fluorogenic), Caspase 4 substrate (fluorogenic). Formula: C33H36F3N5O10. MW: 719.7. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: -20°C. Use/Stability: Solid product is stable for 1 year in desiccator when stored at room temperature. Stock solutions are stable for 6 – 8 months when stored at –20°C. Keep sealed after removing from the freezer until its temperature equilibrates with room temperature. Handling: Keep cool and dry.