Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-YVAD-AFC | 219137-85-6 | 99.9% | 719.66 | 1 MG
SDP

Catalog No. 5000424464
Encompass_Preferred

Ac-YVAD-AFC is a fluorometric peptide substrate of caspase-1. It is utilized in biochemical assays, reacting to produce a fluorescent signal upon enzymatic cleavage. This substrate features specific excitation and emission characteristics for detection.

  • Purity: 99.9%
  • Molecular weight: 719.66
  • Chemical formula: C₃₃H₃₆F₃N₅O₁₀
  • CAS number: 219137-85-6
  • Appearance: Solid, white to off-white
  • Solubility: 100 mg/mL in DMSO
  • Target: Fluorescent Dye

Catalog No. 50-004-24464 Supplier Medchemexpress LLC Supplier No. HYP23771MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-YVAD-AFC is a fluorometric peptide substrate of caspase-1. It is utilized in biochemical assays, reacting to produce a fluorescent signal upon enzymatic cleavage. This substrate features specific excitation and emission characteristics for detection.

  • Purity: 99.9%
  • Molecular weight: 719.66
  • Chemical formula: C₃₃H₃₆F₃N₅O₁₀
  • CAS number: 219137-85-6
  • Appearance: Solid, white to off-white
  • Solubility: 100 mg/mL in DMSO
  • Target: Fluorescent Dye

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.