Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ac-YVAD-AFC | 219137-85-6 | 99.9% | 719.66 | 10 MG
SDP

Catalog No. 5000424465
Encompass_Preferred

Ac-YVAD-AFC is a fluorometric peptide substrate of caspase-1, with an excitation of 400 nm and an emission of 505 nm. It acts as a fluorescent dye target and is classified under the 'Others' pathway.

  • Purity of 99.9%
  • Fluorometric peptide substrate for caspase-1
  • Molecular weight of 719.66
  • Appears as a white to off-white solid
  • Excitation at 380 nm and emission at 460 nm
  • Stable as powder for up to 2 years at -80°C
  • Stable in solvent for up to 6 months at -80°C
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic)

Catalog No. 50-004-24465 Supplier Medchemexpress LLC Supplier No. HYP237710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ac-YVAD-AFC is a fluorometric peptide substrate of caspase-1, with an excitation of 400 nm and an emission of 505 nm. It acts as a fluorescent dye target and is classified under the 'Others' pathway.

  • Purity of 99.9%
  • Fluorometric peptide substrate for caspase-1
  • Molecular weight of 719.66
  • Appears as a white to off-white solid
  • Excitation at 380 nm and emission at 460 nm
  • Stable as powder for up to 2 years at -80°C
  • Stable in solvent for up to 6 months at -80°C
  • Soluble in DMSO at 100 mg/mL (requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.