Learn More
Enzo Life Sciences Ac-YVAD-AFC (5mg)
SDP
Catalog No. 502008319
Encompass_Preferred
Fluorogenic substrate for caspase-1 and -4. Alternative name: Caspase 1 substrate (fluorogenic), Caspase 4 substrate (fluorogenic). Formula: C33H36F3N5O10. MW: 719.7. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8319 Supplier Enzo Life Sciences Supplier No. ALX260108M005
Fluorogenic substrate for caspase-1 and -4. Alternative name: Caspase 1 substrate (fluorogenic), Caspase 4 substrate (fluorogenic). Formula: C33H36F3N5O10. MW: 719.7. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.