Learn More
Cayman Chemical Ac-YVAD-AFC 5mg
SDP
Catalog No. 502979784
Encompass_Preferred
A fluorogenic substrate whose amino acids YVAD have been shown to be a preferred cleavage site for caspase-1 and -4
Catalog No. 50-297-9784 Supplier Cayman Chemical Supplier No. 175915 MG
A fluorogenic substrate whose amino acids YVAD have been shown to be a preferred cleavage site for caspase-1 and -4