Learn More
CPC Scientific Ac-Tyr-Val-Ala-Asp-AMC 1MG
SDP
Catalog No. 502108938
Encompass
ONE-LETTER SEQUENCE: Ac-YVAD-AMCMOLECULAR FORMULA: C33H39N5O10MOLECULAR WEIGHT:665.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [149231-65-2], RESEARCH AREA: ApoptosisREFERENCES: S. H. Inayat-Hussain et al., Hepatology, 25, 1516 (1997); N.A. Thornberry et al., Nature, 356, 768 (1992)
Catalog No. 50-210-8938 Supplier CPC Scientific Supplier No. CASP010A
ONE-LETTER SEQUENCE: Ac-YVAD-AMCMOLECULAR FORMULA: C33H39N5O10MOLECULAR WEIGHT:665.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [149231-65-2], RESEARCH AREA: ApoptosisREFERENCES: S. H. Inayat-Hussain et al., Hepatology, 25, 1516 (1997); N.A. Thornberry et al., Nature, 356, 768 (1992)