Learn More
Medchemexpress LLC Ac-YVAD-AMC | 149231-65-2 | MFCD00171369 | 99.6% | 665.69 | 10 MG
SDP
Catalog No. 5000397453
Encompass_Preferred
Ac-YVAD-AMC is a synthetic tetrapeptide substrate and a caspase inhibitor. It is utilized in research for inhibiting bacteria-induced cell death within hypersensitive response (HR) cells.
  • Cleaved by Caspase-1, releasing fluorogenic AMC.
Catalog No. 50-003-97453 Supplier Medchemexpress LLC Supplier No. HYP271710MG
Ac-YVAD-AMC is a synthetic tetrapeptide substrate and a caspase inhibitor. It is utilized in research for inhibiting bacteria-induced cell death within hypersensitive response (HR) cells.
  • Cleaved by Caspase-1, releasing fluorogenic AMC.