Learn More
Enzo Life Sciences Ac-YVAD-2,6-dimethylbenzoyloxymethylketone (5mg)
SDP
Catalog No. 502008223
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4. Alternative name: Caspase 1 inhibitor, Ac-Tyr-Val-Ala-Asp-2,6-dimethylbenzoyloxymethylketone. Formula: C33H42N4O10. MW: 654.7. Purity: ≥95% (HPLC). Appearance: White powder. Peptide content: 78-99%. Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8223 Supplier Enzo Life Sciences Supplier No. ALX260016M005
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4. Alternative name: Caspase 1 inhibitor, Ac-Tyr-Val-Ala-Asp-2,6-dimethylbenzoyloxymethylketone. Formula: C33H42N4O10. MW: 654.7. Purity: ≥95% (HPLC). Appearance: White powder. Peptide content: 78-99%. Solubility: Soluble in DMSO or methanol. Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. Handling: Keep cool and dry.