Learn More
CPC Scientific Ac-Tyr-Val-Ala-Asp-CHO 1MG
SDP
Supplier:  CPC Scientific CASP011A
Encompass
Tetrapeptide aldehyde, a potent, specific, reversible inhibitor of caspase-1 (ICE) that displays a Ki of 0.2 ±0.1 nM for recombinant human caspase-1.ONE-LETTER SEQUENCE: Ac-YVAD-CHOMOLECULAR FORMULA: C23H32N4O8MOLECULAR WEIGHT:492.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [143313-51-3], RESEARCH AREA: ApoptosisREFERENCES: N.A. Thornberry et al., Nature 356, 768 (1992); S.M. Molineaux et al., Proc. Natl. Acad. Sci. USA, 90, 1809 (1993); N.P.C. Walker et al., Cell, 78, 343 (1994)
Catalog No. 50-210-8940