Learn More
Medchemexpress LLC Ac-YVAD-CHO | 143313-51-3 | 98.2% | 1 MG
SDP
Catalog No. 5000232306
Encompass_Preferred
Ac-YVAD-CHO is a potent, reversible tetrapeptide inhibitor of interleukin-1β converting enzyme (ICE/caspase-1), with reported Ki values of 3.0 nM (mouse) and 0.76 nM (human). It is supplied as a white to off-white solid of high purity for research use in biochemical and cell-based assays targeting caspase-1/IL-1β processing.
  • Potent, reversible inhibitor of caspase-1 (ICE).
  • Low nanomolar Ki for mouse and human enzymes.
  • Suppresses production of mature IL-1β in biochemical assays.
  • High purity suitable for analytical and cell-based studies.
  • Available in small milligram sizes for laboratory research.
  • Store as a powder at -80°C or -20°C to preserve stability.
Catalog No. 50-002-32306 Supplier Medchemexpress LLC Supplier No. HY1200191MG
Ac-YVAD-CHO is a potent, reversible tetrapeptide inhibitor of interleukin-1β converting enzyme (ICE/caspase-1), with reported Ki values of 3.0 nM (mouse) and 0.76 nM (human). It is supplied as a white to off-white solid of high purity for research use in biochemical and cell-based assays targeting caspase-1/IL-1β processing.
  • Potent, reversible inhibitor of caspase-1 (ICE).
  • Low nanomolar Ki for mouse and human enzymes.
  • Suppresses production of mature IL-1β in biochemical assays.
  • High purity suitable for analytical and cell-based studies.
  • Available in small milligram sizes for laboratory research.
  • Store as a powder at -80°C or -20°C to preserve stability.