Learn More
Medchemexpress LLC Ac-YVAD-cmk | 178603-78-6 | 98.9% | 540.99 | 5 MG
SDP
Catalog No. 5000364185
Encompass_Preferred
Ac-YVAD-cmk (Caspase-1 Inhibitor II) is a selective caspase-1 (IL-1beta converting enzyme, ICE) inhibitor. It exhibits neuroprotective and anti-inflammatory effects by effectively suppressing the expression of IL-1β and IL-18, and inhibits pyroptosis in various diseases.
  • Selective caspase-1 (ICE) inhibitor
  • Exhibits neuroprotective effects
  • Offers anti-inflammatory properties
  • Suppresses IL-1β and IL-18 expression
  • Inhibits pyroptosis
  • For research use only
Catalog No. 50-003-64185 Supplier Medchemexpress LLC Supplier No. 5MGHY16990
Ac-YVAD-cmk (Caspase-1 Inhibitor II) is a selective caspase-1 (IL-1beta converting enzyme, ICE) inhibitor. It exhibits neuroprotective and anti-inflammatory effects by effectively suppressing the expression of IL-1β and IL-18, and inhibits pyroptosis in various diseases.
  • Selective caspase-1 (ICE) inhibitor
  • Exhibits neuroprotective effects
  • Offers anti-inflammatory properties
  • Suppresses IL-1β and IL-18 expression
  • Inhibits pyroptosis
  • For research use only