Learn More
Medchemexpress LLC Ac-YVAD-cmk | 178603-78-6 | MFCD00237124 | 98.9% | 540.99 g/mol | C24H33ClN4O8 | 10 MG
SDP
Catalog No. 5000294944
Encompass_Preferred
Ac-YVAD-cmk is a peptide-based, irreversible inhibitor of caspase-1 (interleukin-1β converting enzyme) used in biochemical and cell-based research to block caspase-1 activity and reduce maturation of proinflammatory cytokines. It is commonly applied in studies of inflammation, pyroptosis, and neuroprotection.
  • Selective inhibition of caspase-1 activity in biochemical assays and cell models.
  • Irreversible peptide-based inhibitor for sustained target engagement.
  • Reduces processing of IL-1β and IL-18 in inflammasome-related research.
  • High supplied purity suitable for research applications.
  • Available in multiple pack sizes and as a ready-to-use DMSO solution.
  • Accompanied by datasheet, SDS, and certificate of analysis for reproducible experiments.
Catalog No. 50-002-94944 Supplier Medchemexpress LLC Supplier No. HY1699010MG
Ac-YVAD-cmk is a peptide-based, irreversible inhibitor of caspase-1 (interleukin-1β converting enzyme) used in biochemical and cell-based research to block caspase-1 activity and reduce maturation of proinflammatory cytokines. It is commonly applied in studies of inflammation, pyroptosis, and neuroprotection.
  • Selective inhibition of caspase-1 activity in biochemical assays and cell models.
  • Irreversible peptide-based inhibitor for sustained target engagement.
  • Reduces processing of IL-1β and IL-18 in inflammasome-related research.
  • High supplied purity suitable for research applications.
  • Available in multiple pack sizes and as a ready-to-use DMSO solution.
  • Accompanied by datasheet, SDS, and certificate of analysis for reproducible experiments.