Learn More
Enzo Life Sciences Ac-YVAD-CMK (5mg). CAS: 178603-78-6
SDP
Catalog No. 502008237
Encompass_Preferred
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4. Inhibits Fas-mediated apoptosis. Alternative name: Caspase-1 inhibitor (chloromethylketone). Formula: C24H33N4O8Cl. MW: 541.0. Purity: ≥96% (HPLC). Solubility: Soluble in DMSO (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8237 Supplier Enzo Life Sciences Supplier No. ALX260028M005
Irreversible, cell permeable inhibitor of caspase-1. Also inhibits caspase-4. Inhibits Fas-mediated apoptosis. Alternative name: Caspase-1 inhibitor (chloromethylketone). Formula: C24H33N4O8Cl. MW: 541.0. Purity: ≥96% (HPLC). Solubility: Soluble in DMSO (50mg/ml). Long Term Storage: -20°C.