Learn More
Selleck Chemical LLC Ac-YVAD-cmk-5mg
SDP
Catalog No. 502389753
Encompass_Preferred
Ac-YVAD-cmk (Caspase-1 Inhibitor II) is a selective IL-1β converting enzyme (ICE) inhibitor with neuroprotective and anti-inflammatory effects, effectively suppresses the expression of IL-1β and IL-18.
Catalog No. 50-238-9753 Supplier Selleck Chemical LLC Supplier No. S97275MG
Ac-YVAD-cmk (Caspase-1 Inhibitor II) is a selective IL-1β converting enzyme (ICE) inhibitor with neuroprotective and anti-inflammatory effects, effectively suppresses the expression of IL-1β and IL-18.