Learn More
TARGETMOL CHEMICALS INC AC-YVAD-CMK 5MG
SDP
Catalog No. 502580188
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Ac-YVAD-CMK (Ac-Tyr-Val-Ala-Asp-CMK) is a selective and irreversible inhibitor of Caspase-1 and inhibits pyroptosis IL-1(beta) and IL-18. Ac-YVAD-CMK shows neuroprotective and anti-inflammatory capacity. purity: 97%
Catalog No. 50-258-0188 Supplier TARGETMOL CHEMICALS INC Supplier No. T363475MG
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Ac-YVAD-CMK (Ac-Tyr-Val-Ala-Asp-CMK) is a selective and irreversible inhibitor of Caspase-1 and inhibits pyroptosis IL-1(beta) and IL-18. Ac-YVAD-CMK shows neuroprotective and anti-inflammatory capacity. purity: 97%