Learn More
Carbosynth LLC. Ac-Tyr-Val-Ala-Asp-MCA [Ac-YVAD-MCA] (5 mg vial), 1ea, MW: 665.69, Molecular formula: C33H39N5O10, CAS# 149231-65-2
SDP
Supplier:  Carbosynth LLC. MAY3161V5MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Synonyms: Acetyl-L-Tyrosyl-L-Valyl-L-Alanyl-L-Aspartic Acid -(4-Methyl-Coumaryl-7-Amide), Ac-Tyr-Val-Ala-Asp-AMC, Ac-YVAD-AMC, Purity: , Grade A, Application: Substrate for Caspase-1, Storage Temperature: -20C, References: N.A. Thornberry, H.G. Bull, J.R. Calaycay, K.T. Chapman, A.D. Howard, M.J. Kostura, D.K. Miller, S.M. Molineaux, J.R. Weidner, J. Aunins, K.O. Elliston, J.M. Ayala, F.J. Casano, J. Chin, G.J.-F. Ding, L.A. Egger, E.P. Gaffney, G. Limjuco, O.C. Palyha, S.M. Raju, A.M. Rolando, J.P. Salley, T.T. Yamin, T.D. Lee, J.E. Shively, M. MacCross, R.A. Mumford, J.A. Schmidt, and M.J. Tocci, Nature, 356, 768 (1992).
Catalog No. 50-168-5938