Learn More
Medchemexpress LLC Ac-YVAD-pNA | 149231-66-3 | 99.9% | 628.63 | 10 MG
SDP
Catalog No. 5000423543
Encompass_Preferred
Ac-YVAD-pNA is a specific substrate for Caspase-1. It can be utilized to detect the activity of Caspase-1, which is a key mediator in inflammatory processes. This product is intended for research use only.
  • Specific substrate for Caspase-1
  • Used to detect Caspase-1 activity
  • Key mediator in inflammatory processes
  • Solid, white to off-white appearance
  • Soluble in DMSO at concentrations ≥ 250 mg/mL
Catalog No. 50-004-23543 Supplier Medchemexpress LLC Supplier No. HYP209110MG
Ac-YVAD-pNA is a specific substrate for Caspase-1. It can be utilized to detect the activity of Caspase-1, which is a key mediator in inflammatory processes. This product is intended for research use only.
  • Specific substrate for Caspase-1
  • Used to detect Caspase-1 activity
  • Key mediator in inflammatory processes
  • Solid, white to off-white appearance
  • Soluble in DMSO at concentrations ≥ 250 mg/mL